missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ MacConkey Agar

Numéro de catalogue. R453802
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
R453802 500 g
R453804 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. R453802 Fournisseur Thermo Scientific™ Code fournisseur RE453802
Il en reste null

The Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.

Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.

Spécifications

Certifications/Compliance Industrial Reference: BAM, COMPF, SMWW, USP.
Description MacConkey Agar
Form Powder
Product Type Agar
Quantity 500 g