Learn More
Thermo Scientific™ Remel™ Lysostaphin Test Kit
Catalog No. R21130
Change view
Click to view available options
Quantity:
25 Tests/Kit
Catalog No. Quantity
R21130 25 Tests/Kit
1 options
Catalog No. R21130 Supplier Thermo Scientific™ Supplier No. R21130
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thermo Scientific™ Remel Lysostaphin Test Kit is used for rapid differentiation of staphylococci from micrococci.
Thermo Scientific™ Remel Lysostaphin Test Kit is used for rapid differentiation of staphylococci from micrococci.
Includes:
1 x 4mL vial phosphate buffer and 1 x 5mL Lysostaphin stock solution
Specifications
Content And Storage 2°C to 8°C
Description Lysostaphin Test Kit
Detectable Analytes Staphylococcus species
Includes 1 x 4mL vial phosphate buffer and 1 x 5mL Lysostaphin stock solution
Quantity 25 Tests/Kit