Learn More
Thermo Scientific™ Letheen Broth
Effectively carry out testing of quaternary ammonium compounds for antimicrobial activity using Letheen Broth.
$646.86 - $646.86
Specifications
Description Letheen Broth
Product Type Microbiology Media
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
R112423
View Documents
Thermo Scientific™
R112423
10 x 99 mL
Case of 10 Each for $646.86
Only null left
Description
Description
Effectively carry out testing of quaternary ammonium compounds for antimicrobial activity using Thermo Scientific™ Remel™ Letheen Broth. This subculture medium was developed for neutralization of quaternary ammonium compounds1. Quisno, Gibby, and Foter discovered that adding lecithin and polysorbate 80 to FDA Broth resulted in a medium that neutralized high concentrations of quaternary ammonium salts2. It is used to recover bacteria from solutions containing residues of disinfectant used in the sanitization of food utensils and equipments. It is recommended by the Association of Official Analytical Chemists (AOAC) for cultivation of microorganisms from cosmetic samples3.
Effectively carry out testing of quaternary ammonium compounds for antimicrobial activity using Letheen Broth.
  • Recommended by AOAC for cultivation of microorganisms from cosmetic samples.
  • Recommended by USP in disinfectant challenge testing (The united states pharmacopoeia/National Formulary 2019, US pharmacopial conventionInc. Rockville, M.D.)
  • Ready to use convenience of prepared media.
This medium contains meat peptone and beef extract that supply nitrogenous compounds and amino acids necessary for the growth of bacteria. Lecithin neutralizes quaternary ammonium compounds and polysorbate 80 neutralizes phenolic compounds, formalin and hexachlorophene.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
Letheen Broth
Microbiology Media
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'