Learn More
Thermo Scientific™ Letheen Broth
Catalog No. R112423
Change view
Click to view available options
Quantity:
10 x 99 mL
Catalog No. Quantity
R112423 10 x 99 mL
1 options
Catalog No. R112423 Supplier Thermo Scientific™ Supplier No. R112423
Effectively carry out testing of quaternary ammonium compounds for antimicrobial activity using Letheen Broth.
Effectively carry out testing of quaternary ammonium compounds for antimicrobial activity using Thermo Scientific™ Remel™ Letheen Broth. This subculture medium was developed for neutralization of quaternary ammonium compounds1. Quisno, Gibby, and Foter discovered that adding lecithin and polysorbate 80 to FDA Broth resulted in a medium that neutralized high concentrations of quaternary ammonium salts2. It is used to recover bacteria from solutions containing residues of disinfectant used in the sanitization of food utensils and equipments. It is recommended by the Association of Official Analytical Chemists (AOAC) for cultivation of microorganisms from cosmetic samples3.
Effectively carry out testing of quaternary ammonium compounds for antimicrobial activity using Letheen Broth.
  • Recommended by AOAC for cultivation of microorganisms from cosmetic samples.
  • Recommended by USP in disinfectant challenge
Specifications
Certifications/Compliance Industrial Reference: AOAC.
Description Letheen Broth
Format WMB Lug
Product Type Prepared Microbiology Tubes
Quantity 10 x 99 mL