Learn More
Thermo Scientific™ Remel™ LB Agar w/Ampicillin (100μg/mL) (Luria-Bertani)
Catalog No. R110204
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
Catalog No. Format
R110204 12mm x 85mm Monoplate
1 options
Catalog No. R110204 Supplier Thermo Scientific™ Supplier No. RE110204
Use for the cultivation and maintenance of Escherichia coli for genetic and molecular studies.
Thermo Scientific™ Remel LB Agar with Ampicillin (100μg/mL) (Luria-Bertani) is used for the cultivation and maintenance of Escherichia coli for genetic and molecular studies.
Specifications
Description LB (Luria-Bertani) Agar w/Ampicillin 100micrograms/mL
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.