missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Kanamycin Disk

Catalog No. R21126
Change view
Click to view available options
Quantity:
25 Disks/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21126 25 Disks/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21126 Supplier Thermo Scientific™ Supplier No. RE21126

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Confirm Gram-stain reactions and presumptively identify anaerobic, Gram-negative bacilli with this ready-to-use, kanamycin-impregnated disk.

Confirm Gram-stain reaction of anaerobes and presumptively identify anaerobic, Gram-negative bacilli with Thermo Scientific™ Remel™ Kanamycin Disk.

In 1971 Sutter and Finegold1 recommended a method to identify anaerobic, Gram-negative bacilli based on the difference in response to an antibiotic. Accordingly, organisms can be grouped based on their characteristic inhibitory patterns with erythromycin, rifampin, colistin, penicillin, kanamycin and vancomycin2,3.

Presumptively identify anaerobic bacteria with Kanamycin disks.3,4

  • Ready to use: Kanamycin-impregnated disk.
  • Easy to interpret: Sensitive organisms exhibit an inhibitory zone greater than 10 mm around the antibiotic disk while resistant organisms exhibit no inhibitory zone or a zone that is less than 10 mm around the disk.

Kanamycin Disk contains 1000 μg kanamycin.

Disks impregnated with erythromycin, rifampin, colistin, penicillin, kanamycin and vancomycin can be used to identify anaerobic, Gram-negative bacilli based on the zone of inhibition they exhibit. Colistin and vancomycin confirm the Gram-stain reaction, which is helpful with the Clostridia that stain Gram-negative. Penicillin, rifampin and kanamycin separate Bacteroides and Fusobacterium.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Content And Storage 2°C to 8°C
Description Kanamycin, 10µg
Detectable Analytes Anaerobic Gram-negative bacilli (Bacteroides and Fusobacterium species)
Format Vial
Quantity 25 Disks/Vial