Learn More
Thermo Scientific™ Remel™ Isolator Wrap™ Sterile Sabauroud Dextrose Agar Contact Plate w/Lecithin, Polysorbate 80
Catalog No. p-4523875
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
R111825 10/Pk.
1 options
Catalog No. R111825 Supplier Thermo Scientific™ Supplier No. RE111825
Only null left
Thermo Scientific™ Remel Isolator Wrap Sterile Sabauroud Dextrose Agar Contact Plate w/Lecithin, Polysorbate 80 is used for the cultivation of yeasts and fungi.
Thermo Scientific™ Remel Isolator Wrap Sterile Sabauroud Dextrose Agar Contact Plate w/Lecithin, Polysorbate 80 is used for the cultivation of yeasts and fungi.
  • Foil Barrier Wrap — impermeable to isolator decontamination process
  • GripFit™ Plates — easy to handle with one hand without lid separating from bottom of plate
  • Irradiated — Sterility Assurance Level 10-6
  • Shrink-wraped with easy open tear strip — reduces breakage and moisture loss
  • Bottom labeling — information stays with sample
  • Extra sterile bag — easy to transport plates to lab without introducing contamination
  • Six month shelf life — less ordering and fewer lot numbers to QC
Specifications
Description Sabouraud Dextrose Agar
Format Foil Barrier Wrap + Bag
Product Type Agar
Quantity 10/Pk.
Not intended for IVD use.