Learn More
Thermo Scientific™ Remel™ Inhibitory Mold Agar
Catalog No. R453592
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R453592 500 g
1 options
Catalog No. R453592 Supplier Thermo Scientific™ Supplier No. RE453592
Only null left
Isolate and cultivate dermatophytes and other pathogenic yeasts and molds using Inhibitory Mold Agar (Dehydrated).
Isolate and cultivate dermatophytes and other pathogenic yeasts and molds using Thermo Scientific™ Remel™ Inhibitory Mold Agar (dehydrated). Ulrich formulated Inhibitory Mold Agar as a general-purpose medium for cultivation of fungi1. It is also recommended for selective isolation of pathogenic yeasts and molds such as Cryptococcus neoformans, Candida spp., and Aspergillus spp. from sources contaminated with bacteria2,3.
Inhibitory Mold Agar is recommended for primary isolation and cultivation of dermatophytes, and other pathogenic fungi.
  • Selective medium: Chloramphenicol inhibits many gram-positive and gram-negative bacteria.
  • Recommended for selective isolation of pathogenic yeasts and molds such as Cryptococcus neoformans, Candida spp., and Aspergillus from sources contaminated with bacteria2,3
This medium contains casein and meat peptones that supply nitrogenous compounds, amino acids, and peptides necessary for the growth of fungi. Dextrose, starch, and dextrin are ready sources of energy. Sodium chloride and metallic salts provide essential electrolytes that help to maintain osmotic equilibrium. Yeast extract acts as a source of vitamins. Chloramphenicol is the antibiotic that acts as a selective agent and inhibits the growth of gram-positive and gram-negative bacteria.
Specifications
Description Inhibitory Mold Agar
Form Powder
Product Type Agar
Quantity 500 g
For Laboratory Use Only