missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Indoxyl Acetate Disk

Catalog No. R21087
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21087 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21087 Supplier Thermo Scientific™ Supplier No. RE21087

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rapidly identify and differentiate Campylobacter, Wolinella, and Helicobacter using Indoxyl Acetate Disk.

Rapidly identify and differentiate Campylobacter, Wolinella, and Helicobacter based on their ability to hydrolyze indoxyl acetate using Thermo Scientific™ Remel™ Indoxyl Acetate Disk. Hodge et al. confirming the findings of Mills et al first found in 1987, reported that Indoxyl Acetate Disk was reliable for differentiation of Campylobacter spp1,2. In 1990, further investigation of the disk by Popovic-Uroic et al. reported the method to be more reliable than nalidixic acid susceptibility for differentiation of Campylobacter spp. and useful in differentiating Wolinella and Helicobacter3.

Rapidly identify and differentiate Campylobacter, Wolinella, and Helicobacter based on their ability to hydrolyze indoxyl acetate using Indoxyl Acetate Disk.

  • Ready to use: Reagent-impregnated disk.
  • Rapid test: Results are obtained within 20 minutes.
  • Easy to identify: A positive result showcases development of blue or blue-green color.

Bacterial esterase releases indoxyl from indoxyl acetate which reacts spontaneously to produce indigo in the presence of oxygen. A total of 981 strains of Campylobacter, Wolinella and Helicobacter were evaluated in separate studies published by Hodge et al and Popovic-Uroic et al2,3.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Content And Storage 2°C to 8°C
Description Indoxyl Acetate Disk used to rapidly identify and differentiate Campylobacter, Wolinella, and Helicobacter based on their ability to hydrolyze indoxyl.
Detectable Analytes Indoxyl
Quantity 25 mL