missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ Horse Defibrinated Blood
Thermo Scientific™ Remel Defibrinated Horse Blood is packaged in various sizes.
$123.89 - $123.89
Specifications
| Species | Horse |
|---|---|
| Form | Liquid |
| Specialty Type | Defibrinated |
| Product Type | Horse Whole Blood |
| Processing | Defibrinated |
Description
Thermo Scientific™ Remel Defibrinated Horse Blood is packaged in various sizes.
Specifications
| Horse | |
| Defibrinated | |
| Defibrinated |
| Liquid | |
| Horse Whole Blood |