missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hippurate Disk

Catalog No. R21085
Change view
Click to view available options
Quantity:
25 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21085 25 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21085 Supplier Thermo Scientific™ Supplier No. RE21085

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rapidly detect the hydrolysis of sodium hippurate by β-hemolytic streptococci, G. vaginalis, and C. jejuni using a reagent-impregnated Hippurate Disk.

Rapidly detect the hydrolysis of sodium hippurate by β-hemolytic streptococci, Gardnerella vaginalis, and Campylobacter jejuni using a reagent-impregnated Thermo Scientific™ Remel™ Hippurate Disk. Hwang and Ederer employed the ninhydrin reagent in a rapid method to detect hippurate hydrolysis by the end product glycine1. Jolly later outlined minimal criteria for identification of G. vaginalis including hippurate hydrolysis2. In 1989, Cacho et al. further evaluated the use of the hippurate disk in the identification of Campylobacter spp3.

Rapidly detect the hydrolysis of sodium hippurate by β-hemolytic streptococci, Gardnerella vaginalis, and Campylobacter jejuni using a reagent-impregnated Hippurate Disk. The hippurate hydrolysis test determines the ability of Group B streptococci, as well as other bacteria, to enzymatically hydrolyze sodium hippurate4,5.

  • Ready to use: Reagent-impregnated disk
  • Rapid test: Results are obtained within 30 minutes.
  • Easy to identify: A condensation reaction occurs with hydrindantin, ammonia, and residual ninhydrin to produce the final purple colored complex for a positive reaction and light gray color for a negative reaction.

Hippuric acid is hydrolyzed by the enzyme hippuricase to glycine and benzoic acid. Ninhydrin evokes a five-step reaction beginning with the deamination of glycine to form hydrindantin (a reduced form of ninhydrin), carbon dioxide, and ammonia.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Content And Storage 2°C to 8°C
Description Hippurate Disk
CE Marker Yes
Format Vial
Packaging Type 1 Kit
Quantity 25 Tests/Kit