Learn More
Thermo Scientific™ Remel™ Haemophilus Test Medium Broth (HTM Broth)
Catalog No. R112380
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
R112380 500 mL
1 options
Catalog No. R112380 Supplier Thermo Scientific™ Supplier No. R112380
Use for antimicrobial minimal inhibitory concentration (MIC) determinations of Haemophilus spp.
Thermo Scientific™ Remel Haemophilus Test Medium Broth (HTM Broth) is for the antimicrobial minimal inhibitory concentration (MIC) determinations of Haemophilus spp.
Specifications
Description Haemophilus Test Medium Broth
Format Clear Square Bottle
Product Type Microbiology Media
Quantity 500 mL
REF R112380 not intended for IVD use.