missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Ferric Chloride

Catalog No. R21218
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21218 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21218 Supplier Thermo Scientific™ Supplier No. R21218

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detect phenylalanine deamination in Enterobacteriaceae with Ferric Chloride.

Detect phenylalanine deamination with Thermo Scientific™ Remel™ Ferric Chloride.Henriksen and Closs reported Proteus could be differentiated from other enteric gram-negative bacilli based on the ability to deaminate phenylalanine into phenylpyruvic acid1-3. This reaction is also associated with Providencia and Morganella spp4,5. 10% Ferric Chloride solution is used to detect phenylpyruvic acid formed in the deamination of phenylalanine6.

Ferric Chloride is used detect phenylalanine deamination in Enterobacteriaceae. Phenylalanine undergoes oxidative deamination to yield phenylpyruvic acid. In presence of 10% Ferric Chloride it chelates with phenylpyruvic acid to form green color.Ready for use: no further preparation is required.Easy to interpret: green color develops within 5 minutes.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Content And Storage 2°C to 8°C
Description Ferric Chloride
Detectable Analytes Phenylpyruvic Acid
Format Bottle
Quantity 25 mL