missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ CTA w/1% Sucrose (Cystine Trypticase Agar)

Catalog No. R060660
Change view
Click to view available options
Quantity:
20 x 4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R060660 20 x 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R060660 Supplier Thermo Scientific™ Supplier No. R060660

Use for determination of fermentation reactions by fastidious microorganisms, such as Neisseria, pneumococci, streptococci and non-spore forming anaerobes.

Thermo Scientific™ Remel CTA with 1% Sucrose (Cystine Trypticase Agar) is used for determination of fermentation reactions by fastidious microorganisms, such as Neisseria, pneumococci, streptococci and non-spore forming anaerobes.

Specifications

Description CTA w/1% Sucrose
Format 15mm x 103mm Tube
Product Type Agar
Quantity 20 x 4 mL