missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cornmeal Agar with Polysorbate 80 with or without Trypan Blue

Catalog No. r01328
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R01328 12mm x 85mm Monoplate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R01328 Supplier Thermo Scientific™ Supplier No. RE01328
Only null left

Cultivate fungi and stimulate chlamydospore production by Candida albicans with this ready-to-use medium.

Cultivate fungi and stimulate chlamydospore production by Candida albicans with Thermo Scientific™ Remel Cornmeal Agar with Polysorbate 80 general purpose medium, with or without Trypan Blue. Polack and Benham demonstrated the use of cornmeal agar for studying differential mycelial characteristics of Candida species1,2. Adding polysorbate 80 to cornmeal agar stimulates chlamydospore formation by Candida albicans3-4.

Cornmeal Agar with Polysorbate 80 with or without trypan blue is a solid medium recommended for use in qualitative procedures as a general-purpose media for cultivating fungi and stimulating chlamydospore production by Candida albicans.

  • General-purpose media: cultivate fungi and stimulate chlamydospore formation by Candida albicans.
  • Easy-to-read: Trypan blue provides a contrasting background for the optimal visualization of chlamydospores.
  • Ready-to-use: the convenience of prepared media.

Cornmeal agar consists of an infusion of cornmeal and agar, which provides the essential nutrients needed to support the growth of fungi and the production of chlamydospores by Candida albicans. The agar is a solidifying agent. Polysorbate 80 reduces surface tension and enhances the development of chlamydospores. Trypan blue is an optimal additive that provides a contrasting background, thereby permitting optimal visualization of chlamydospores.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Description Cornmeal Agar w/Tween™ 80
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.