Learn More
Thermo Scientific™ Remel™ Contact Plate Sterile Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80 (pH 5.6) (Irradiated)
Catalog No. R111806
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R111806 100/Pk.
R111805 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R111806 Supplier Thermo Scientific™ Supplier No. RE111806
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thermo Scientific™ Remel Contact Plate Sterile Sabouraud Dextrose Agar with Lecithin, Polysorbate 80 (pH 5.6, Irradiated) is used for cultivation of yeast and fungi.
Thermo Scientific™ Remel Contact Plate Sterile Sabouraud Dextrose Agar with Lecithin, Polysorbate 80 (pH 5.6) (Irradiated) is used for the cultivation of yeast and fungi.
  • Contact Plates Sterile are double bagged plus a 3rd bag is enclosed for customer use (Double + Bag)
  • These products are manufactured in a clean room and validated sterile by gamma radiation
  • The Contact Plates Sterile listed are made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
Specifications
Description Contact Plates; Sabouraud Dextrose Agar w/Lecithin; Gamma irradiated
Format Contact - Double Bag
Product Type Agar Plate
Quantity 100/Pk.
Not intended for IVD use.