Learn More
Thermo Scientific™ Remel™ Contact I Rose Bengal Agar
Catalog No. R111042
Catalog No. R111042 Supplier Thermo Scientific™ Supplier No. RE111042
Thermo Scientific™ Remel Contact I Rose Bengal Agar is used for selective isolation and enumeration of yeast, and fungi from environmental samples and food.
Thermo Scientific™ Remel Contact I Rose Bengal Agar is used for selective isolation and enumeration of yeast, and fungi from environmental samples and food.
Specifications
Description Contact I Rose Bengal Agar w/chloramphenicol
Format Contact - Single Bag
Product Type Agar Plate
Quantity 20/Pk.