Learn More
Thermo Scientific™ Coagulase Plasma
Detect coagulase enzyme in staphylococci with Coagulase Plasma.
Supplier:  Thermo Scientific™ R21052
 View more versions of this product
Catalog No. R21052
Only null left
Description
Description
Detect coagulase enzyme in staphylococci with Thermo Scientific™ Remel™ Coagulase Plasma. The enzyme coagulase acts on a constituent of rabbit plasma to produce a thrombin-like substance which activates fibrinogen to form fibrin clot. In 1940, Fairbrother and Chapman et al. reported pathogenic staphylococci can be identified based on the ability to coagulate plasma.1,2 Chapman found rabbit plasma to be superior to other types of plasma in terms of clotting activity.3 Bayliss and Hall recommended replacing the citrate anticoagulant with EDTA to avoid false-positive clots.4
  • Rabbit plasma with EDTA
  • Lyophilized
Specifications
Chemical Identifiers
Coagulase Plasma
Staphylococcus species
Enzyme Immunoassay Detection
Yes
Coagulase Plasma
Specifications
Coagulase Plasma
Staphylococcus species
Enzyme Immunoassay Detection
Yes
Coagulase Plasma
25 mL
Product Suggestions
Product Suggestions