Learn More
Thermo Scientific™ Coagulase Plasma
Catalog No. R21051
Change view
Click to view available options
Quantity:
5 mL
15 mL
25 mL
6 x 5 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21051 15 mL
R21060 6 x 5 mL
R21050 5 mL
R21052 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R21051 Supplier Thermo Scientific™ Supplier No. R21051
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Detect coagulase enzyme in staphylococci with Coagulase Plasma.
Detect coagulase enzyme in staphylococci with Thermo Scientific™ Remel™ Coagulase Plasma. The enzyme coagulase acts on a constituent of rabbit plasma to produce a thrombin-like substance which activates fibrinogen to form fibrin clot. In 1940, Fairbrother and Chapman et al. reported pathogenic staphylococci can be identified based on the ability to coagulate plasma.1,2 Chapman found rabbit plasma to be superior to other types of plasma in terms of clotting activity.3 Bayliss and Hall recommended replacing the citrate anticoagulant with EDTA to avoid false-positive clots.4
  • Rabbit plasma with EDTA
  • Lyophilized
Specifications
Description Coagulase Plasma
CE Marker Yes
Detectable Analytes Staphylococcus species
Type Coagulase Plasma
For Use With (Application) Enzyme Immunoassay Detection
Quantity 15 mL