Learn More
Thermo Scientific™ Remel™ Charcoal Blood Agar w/cephalexin (Regan-Lowe Medium)
Description
Thermo Scientific™ Remel Charcoal Blood Agar w/cephalexin (Regan-Lowe Medium) is used for selective isolation of Bordetella pertussis and Bordetella parapertussis.
Specifications
Specifications
| Format | Monoplate |
| Media Type | Charcoal Blood Agar |
| Product Type | Agar |
| Includes | Cephalexin |
| Quantity | 10/Pk. |