missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Campy Thioglycollate Medium (w/.16% Agar)

Catalog No. R060372
Change view
Click to view available options
Quantity:
20 x 9 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R060372 20 x 9 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R060372 Supplier Thermo Scientific™ Supplier No. NRE2430

Used as a holding medium for samples prior to inoculation of media for recovery of Campylobacter spp.

Thermo Scientific™ Remel Campy Thioglycollate Medium (w/.16% agar, trimethoprim, vancomycin, polymyxin B, cephalothin, amphotericin B) is used as a holding medium for samples prior to inoculation of media for recovery of Campylobacter spp.

Specifications

Description Campy Thioglycollate Medium
Format 15mm x 103mm Tube
Product Type Agar Tube
Quantity 20 x 9 mL