missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Butterfield's Phosphate Buffer

Catalog No. p-4522922
Change view
Click to view available options
Quantity:
1000 mL
10 x 90 mL
10 x 100 mL
Format:
PB -1000mL
PB -100mL
PB -90mL
Packaging Type:
10/Pk.
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Packaging Type
R112519 10 x 100 mL PB -100mL 10/Pk.
R112520 1000 mL PB -1000mL Each
R112526 10 x 90 mL PB -90mL 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R112519 Supplier Thermo Scientific™ Supplier No. RE112519
Only null left

Thermo Scientific™ Remel Butterfield's Phosphate Buffer (pH 7.2) is used as a diluent in microbial limit testing of pharmaceutical products and food testing.

Thermo Scientific™ Remel Butterfield's Phosphate Buffer (pH 7.2) is used as a diluent in microbial limit testing of pharmaceutical products and food testing.

Not intended for IVD use.

Specifications

Certifications/Compliance Industrial Reference: BAM, USDA, USP.
Description Phosphate Buffer, Butterfield's
Format PB -100mL
Product Type Buffer Solution
Packaging Type 10/Pk.
Quantity 10 x 100 mL