missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ Butterfield's with 0.1% Peptone Phosphate Buffer
Thermo Scientific™ Remel Butterfield's with 0.1% Peptone Phosphate Buffer is for use as a diluent in microbial limits testing.
$1529.63 - $1529.63
Specifications
| Media Type | Phosphate Buffer-Butterfield's |
|---|---|
| Product Type | Buffer Solution |
| Includes | 0.1% Peptone |
| Quantity | 100 x 9 mL |
| Catalog Number | Mfr. No. | Format | Media Type | Includes | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Format | Media Type | Includes | Price | Quantity | |||
|
R07601
|
Thermo Scientific™
RE07601 |
16mm x 125mm Tube | Phosphate Buffer-Butterfield's | 0.1% Peptone |
Case of 100 Each for $1,529.63
|
|
|||
Description
Thermo Scientific™ Remel Butterfield's with 0.1% Peptone Phosphate Buffer is for use as a diluent in microbial limits testing.
Specifications
| Phosphate Buffer-Butterfield's | |
| Buffer Solution | |
| 0.1% Peptone | |
| 100 x 9 mL |