missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Butterfield's with 0.1% Peptone Phosphate Buffer

Catalog No. R07601
Change view
Click to view available options
Format:
16mm x 125mm Tube
Media Type:
Phosphate Buffer-Butterfield's
Includes:
0.1% Peptone
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Media Type Includes
R07601 16mm x 125mm Tube Phosphate Buffer-Butterfield's 0.1% Peptone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R07601 Supplier Thermo Scientific™ Supplier No. RE07601

Thermo Scientific™ Remel Butterfield's with 0.1% Peptone Phosphate Buffer is for use as a diluent in microbial limits testing.

Thermo Scientific™ Remel Butterfield's with 0.1% Peptone Phosphate Buffer is for use as a diluent in microbial limits testing.

Specifications

Format 16mm x 125mm Tube
Media Type Phosphate Buffer-Butterfield's
Product Type Buffer Solution
Includes 0.1% Peptone
Quantity 100 x 9 mL