Learn More
Thermo Scientific™ Brilliant Green Agar
Description
Selectively isolate and differentiate Salmonella species other than Salmonella typhi from food and other materials with this selective medium.
Selectively isolate and differentiate Salmonella species other than Salmonella typhi from food and other materials with Thermo Scientific™ Remel™ Brilliant Green Agar. The medium is recommended by the American Public Health (APHA) for the recovery of Salmonella from food products1.
Salmonella infection (salmonellosis) is a common bacterial disease that affects the intestinal tract. Salmonella bacteria typically live in animal and human intestines and are shed through feces. Humans become infected most frequently through contaminated water or food.
Use Brilliant Green Agar for isolation of salmonellae other than Salmonella typhi.
- Selective: brilliant green in the medium inhibits the growth of non-target organisms.
- Easy-to-read: Salmonella colonies are easy to read as they range from reddish to pink to nearly white in color with red zone.
- Corresponds to the medium recommended by the APHA1
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: SMWW, USDA, USP. |
| Description | Brilliant Green Agar |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
Not intended for IVD use.