Learn More
Thermo Scientific™ Remel™ Biplates: Anaerobic Reducible through Levine Agars
Catalog No. R02065
Change view
Click to view available options
Quantity:
10/Pk.
Media Type:
Columbia CNA/MacConkey Agar
Catalog No. Quantity Media Type
R02065 10/Pk. Columbia CNA/MacConkey Agar
1 options
Catalog No. R02065 Supplier Thermo Scientific™ Supplier No. RE02065
Perform screening and isolation in the same 12 x 85mm dish, divided into two sections to hold either the same media on both sides or two media, one on each side.
Perform screening and isolation in the same 12 x 85mm dish using Thermo Scientific™ Remel™ Biplates, divided into two sections to hold either the same media on both sides or two media, one on each side.
Specifications
Format Biplate
Media Type Columbia CNA/MacConkey Agar
Product Type Agar Plate
Quantity 10/Pk.