missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ BHI Broth w/6.5% NaCl

Catalog No. p-4523786
Change view
Click to view available options
Quantity:
20 x 4.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R060286 20 x 4.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R060286 Supplier Thermo Scientific™ Supplier No. RE060286
Only null left

Thermo Scientific™ Remel BHI Broth with 6.5% NaCl (Brain Heart Infusion Broth) is for the differentiation of enterococci from nonenterococcal group D streptococci.

Thermo Scientific™ Remel BHI Broth with 6.5% NaCl (Brain Heart Infusion Broth) is for the differentiation of enterococci from nonenterococcal group D streptococci.

Specifications

Description BHI Broth w/6.5% NaCl
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 4.5 mL