missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ BactiDrop™ Ferric Chloride

Catalog No. R21514
Change view
Click to view available options
Quantity:
50 x 0.75 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21514 50 x 0.75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21514 Supplier Thermo Scientific™ Supplier No. RE21514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use BactiDrop™ Ferric Chloride (10%) reagent for detecting phenylalanine deamination in the phenylalanine test.

Detect deamination of phenylalanine using Thermo Scientific™ Remel™ BactiDrop™ Ferric Chloride (10%) reagent. Henecka and Houben-Weyl demonstrated that when testing for the presence of phenylpyruvic acid using an aqueous solution of ferric chloride, a green color reaction is produced1,2. This biochemical test is useful for differentiating Enterobacteriaceae. Most members of the Proteus and Providencia groups give a positive phenylalanine test reaction3,4.

The ability to deaminate phenylalanine to phenylpyruvic acid is measured by the phenylalanine test. Ferric chloride acts as a chelating agent that reacts with phenylpyruvic acid and forms a green color5.Ready-to-use: Liquid reagentRapid test: Visible color change within 5 minutes of addition of reagentEasy to interpret:Positive Test - Green color development within 5 minutesNegative Test - No color developmentBactiDrop™ Ferric Chloride (10%) contains Ferric Chloride,6H2O in hydrochloric acid and demineralized water and is supplied as 0.75mL ampules.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Content And Storage 20°C to 25°C
Description BactiDrop Ferric Chloride
CE Marker Yes
Quantity 50 x 0.75 mL