Learn More
Thermo Scientific™ Remel™ Anaerobic Blood Agar, Monoplate
Catalog No. R01040
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
R01040 10/Pk.
1 options
Catalog No. R01040 Supplier Thermo Scientific™ Supplier No. RE01040
Isolate and cultivate anaerobic organisms, including fastidious strains with this ready-to-use medium.
Isolate and cultivate anaerobic organisms, including fastidious strains with Thermo Scientific™ Remel™ Anaerobic Blood Agar ready prepared plates. Anaerobic Blood Agar serves as a nutritious medium supporting good growth and typical pigmentation of fastidious and slow growing anaerobes, as well as other anaerobes of clinical importance.
Anaerobe Blood Agar contains peptones which supply nitrogenous substances and amino acids necessary for the growth of anaerobic bacteria. Yeast extract provides B-complex vitamins and serves as a growth enhancer. Hemin, vitamin K, and sheep blood stimulate the growth of anaerobes. Sodium chloride is a source of essential electrolytes and maintains osmotic equilibrium.
Specifications
Description Anaerobic Blood Agar
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.