missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Agglutinating Sera, Salmonella O

Catalog No. R30957101
Change view
Click to view available options
Description:
Factor 4 (group B )
Factor 6, 7 (group C1)
Factor 8 (group C2)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
R30957101 Factor 6, 7 (group C1)
R30957201 Factor 8 (group C2)
R30956901 Factor 4 (group B )
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R30957101 Supplier Thermo Scientific™ Supplier No. RE30957101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Serologically identify somatic O antigens of Salmonella strains by slide agglutination test using Agglutinating Sera, Salmonella O.

Serologically identify somatic O antigens of Salmonella strains by slide agglutination test for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O. The sera are absorbed to remove agglutinins for other Salmonella antigens and the paracolon α agglutinin. Salmonella Somatic Agglutinating Sera (ZC series) are intended for the identification of O antigens.

  • Select sera also for use as positive controls for stained Salmonella suspensions

Specifications

Control Sets Positive
Description Factor 6, 7 (group C1)
Detectable Analytes Salmonella O Antigen
Type Agglutinating Sera
Organism Group Group C1
Packaging Type 2 mL/Vial
Quantity 2 mL
Target Organism Class Salmonella O