missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Agglutinating Sera, Salmonella O/Vi

Catalog No. R30957401
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R30957401 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R30957401 Supplier Thermo Scientific™ Supplier No. R30957401

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Serologically identify somatic O/Vi antigens of Salmonella strains by slide and/or tube agglutination test using Agglutinating Sera, Salmonella O/Vi.

Serologically identify somatic O/Vi antigens of Salmonella strains by slide and/or tube agglutination test for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O/Vi. The sera are absorbed to remove agglutinins for other Salmonella antigens and the paracolon α agglutinin. Salmonella Somatic Agglutinating Sera (ZC series) are intended for the identification of O/Vi antigens.

Serological tests are based on the fact that antibodies in serum, produced in response to exposure to bacterial antigens, will agglutinate with bacteria carrying homologous antigens.

The genus Salmonella is classified in the Kauffmann-White Scheme into serotypes according to combinations of somatic (O) and flagellar (H) antigens, which are identified by agglutination tests1. Agglutinating sera should be used in confirmatory tests but may also be used with appropriate caution in screening tests2. These simple tests rapidly detect specific bacterial antigens for the identification, grouping and/or typing of Salmonella and other food-borne microorganisms. This incorporates agglutinating sera for the identification somatic O/Vi antigens of Salmonella (groups A-S), allowing the fast, accurate and reliable identification of Salmonella strains.

  • Ready-to-use: Liquid reagents in 2mL vials
  • Rapid: Results within 1 minute.
  • Easy-to-interpret: Visible agglutination indicates a positive result
  • Less sample required

Each kit contains one bottle of antisera (2mL) preserved either with 0.5% phenol or 0.1% sodium azide. The sera are produced in rabbits.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Control Sets Positive
Description Salmonella O/Vi
Detectable Analytes Salmonella O Antigen
Type Agglutinating Sera
Material Polyvalent
Packaging Type 2 mL/Vial
Quantity 2 mL
Target Organism Class Salmonella O/Vi