missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O Polyvalent

Catalog No. R30858201
Change view
Click to view available options
Description:
Polyvalent (group A - G)
Polyvalent (group A - S)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
R30858201 Polyvalent (group A - S)
R30858101 Polyvalent (group A - G)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R30858201 Supplier Thermo Scientific™ Supplier No. RE30858201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.

Remel Agglutinating Sera, Salmonella O is used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.

  • Select sera also for use as positive controls for stained Salmonella suspensions

Specifications

Control Sets Positive
Description Polyvalent (group A - S)
Detectable Analytes Salmonella O Antigen
Type Agglutinating Sera
Material Polyvalent
Organism Group Group A-S
Packaging Type 2 mL/Vial
Quantity 2 mL
Target Organism Class Salmonella O