Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Supplier: Thermo Scientific™ RE30858501
Description
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
- Select sera also for use as positive controls for stained Salmonella suspensions
Chemical Identifiers
| Positive | |
| Agglutinating Sera | |
| 2 mL |
| Salmonella H Antigen | |
| 2 mL/Vial |
Specifications
| Positive | |
| Salmonella H Antigen | |
| Polyvalent | |
| 2 mL |
| Polyvalent, Phases 1 and 2 | |
| Agglutinating Sera | |
| 2 mL/Vial | |
| Salmonella H |