Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Catalog No. R30959801
Change view
Click to view available options
Description:
Polyvalent, Phases 1 and 2
Rapid Diagnostic 1 (b, d, E, r)
Rapid Diagnostic 2 (b, E, k, L)
Rapid Diagnostic 3 (d, E, G, k)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
R30959801 Rapid Diagnostic 1 (b, d, E, r)
R30858501 Polyvalent, Phases 1 and 2
R30160101 Rapid Diagnostic 3 (d, E, G, k)
R30959901 Rapid Diagnostic 2 (b, E, k, L)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R30959801 Supplier Thermo Scientific™ Supplier No. RE30959801
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
Specifications
Control Sets Positive
Description Rapid Diagnostic 1 (b, d, E, r)
Detectable Analytes Salmonella H Antigen
Type Agglutinating Sera
Packaging Type 2 mL/Vial
Quantity 2 mL
Target Organism Class Salmonella H