Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Supplier:  Thermo Scientific™ RE30160101
 View more versions of this product
Catalog No. R30160101
Only null left
Description
Description
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
Specifications
Chemical Identifiers
Positive
Agglutinating Sera
2 mL
Salmonella H Antigen
2 mL/Vial
Specifications
Positive
Salmonella H Antigen
2 mL/Vial
Salmonella H
Rapid Diagnostic 3 (d, E, G, k)
Agglutinating Sera
2 mL
Product Suggestions
Product Suggestions