missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ 2% Bile Solubility Reagent

Catalog No. R21209
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21209 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21209 Supplier Thermo Scientific™ Supplier No. R21209

Use for differentiation of S. pneumoniae from other alpha-hemolytic streptococci using the Spot Test or Tube Test.

Thermo Scientific™ Remel 2% Bile Solubility Reagent is for differentiation of S. pneumoniae from other alpha-hemolytic streptococci using the Spot Test or Tube Test.

Specifications

Description Bile Solubility Reagent, 2%
Detectable Analytes S. pneumoniae
Format Bottle
Quantity 25 mL