Learn More
Pharmacal Research Labs Pharmacal Research Labs Release Glass Detergent, Highly Concentrated, Non-Foaming, 5 gal DFS Item
Supplier:  Pharmacal Research Labs 04525F
The RELEASE Glass Detergent by PHARMACAL RESEARCH LABS is a highly concentrated alkaline detergent designed for thorough cleaning in lab environments. It effectively removes a variety of soils, leaving glassware spot-free. With non-foaming and free-rinsing properties, it is suitable for mechanical glassware washers and ultrasonic cleaners. Its thermal stability above 180F ensures optimal performance under rigorous conditions, making it ideal for a range of cleaning applications.
Catalog No. NC9733700
Only null left
Explore available promotions