missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BINDER™ Reinforced Inner Chamber
Reinforced inner chamber for climate chambers and cooling incubation chambers.
Supplier: BINDER™ 80122526
Description
- Reinforced inner chamber for bulky and heavy samples.
- Weight capacity ranged from 100 to 200 kg, depending on model.
Specifications
| KBF PRO 470 | |
| Stainless Steel | |
| Chamber Rack | |
| 180 kg |