missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Reinforced Inner Chamber

For Use With: KMF 240

Supplier:  BINDER™ 80120624

Catalog No. 08111236

Only null left
Explore available promotions

Description

Description

  • Max. total load of 85 to 350 kg
Specifications

Specifications

KMF 240
Max. load 250 kg
Stainless Steel
Reinforced Chamber
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions