missing translation for 'onlineSavingsMsg'
Learn More

Reinecke Salt, ACS Reagent

Catalog No. p-3734724
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423330250 25 g Glass bottle
AC423331000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC423330250 Supplier Thermo Scientific Chemicals Supplier No. 423330250
Only null left

CAS: 13573-16-5 | C4H10CrN7S4·H2O | 354.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13573-16-5
Molecular Formula C4H10CrN7S4·H2O
Molecular Weight (g/mol) 354.46
MDL Number MFCD00150925
Synonym Ammonium reineckate, Ammonium tetrathiocyanatodiamminechromate

Specifications

Melting Point 270.0°C
Color Red
Physical Form Crystalline Powder
Quantity 25 g
Linear Formula NH4(Cr(NH3)2(SCN)4)·H2O
Merck Index 15, 8248
Solubility Information Solubility in water: soluble in hot water. Other solubilities: insoluble in deluted hydrochloric acid,soluble in ethanol
Formula Weight 354.46
Grade ACS Reagent
Infrared Spectrum Authentic
Packaging Glass bottle
Sensitivity Passes Test
Chemical Name or Material Reinecke salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 237-003-3

RUO – Research Use Only