missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reinecke Salt, ACS Reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 270.0°C |
| Color | Red |
| Physical Form | Crystalline Powder |
| Quantity | 100 g |
| Linear Formula | NH4(Cr(NH3)2(SCN)4)·H2O |
| Merck Index | 15, 8248 |
| Solubility Information | Solubility in water: soluble in hot water. Other solubilities: insoluble in deluted hydrochloric acid,soluble in ethanol |
| Formula Weight | 354.46 |
| Grade | ACS Reagent |
| Infrared Spectrum | Authentic |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF |
| EINECSNumber | 237-003-3 |
RUO – Research Use Only