missing translation for 'onlineSavingsMsg'
Learn More

Reinecke Salt, ACS Reagent, ≥93.0%, Honeywell Fluka™

Catalog No. p-7120748
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045031 25 g Glass Bottle
60045030 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60045031 Supplier Honeywell Chemicals Supplier No. 18368725G
Only null left

ACS reagent,≥93.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 13573-16-5
MDL Number MFCD00066638

Specifications

Melting Point 272°C
Physical Form Solid
Quantity 25 g
Linear Formula NH4[Cr(NH3)2(SCN)4]
UN Number NONH for all modes of transport
Formula Weight 336.43g/mol
Percent Purity ≥93.0%
Grade ACS Reagent
Packaging Glass Bottle
Chemical Name or Material Reinecke Salt
Hazard Statement P261-P280-P301 + P312 + P330
Precautionary Statement H302 + H312 + H332
EINECSNumber 237-003-3