Learn More
Reichert™ Industrial Fluid Testing Refractometers
Designed specifically for checking ratio or concentration of water-soluble industrial fluids
Supplier:  Reichert™ 13754000
Catalog No. RE10440
Only null left
Includes:
  • Durable plastic carrying case
  • Sample application pipets
  • Calibration adjustment wrench
Description
Description
Portable testers are ideal for applications including metalworking coolants and lubricants, heat-treating fluids, hydraulic fluids, firefighting foam and plating baths.
  • Automatic temperature compensation provides accurate readings without correction tables or temperature measurement
  • Temperature-compensation range, 15° to 38°C (59° to 100°F)
  • Large, easy-to-read scales and depth of field
  • Portable: no batteries or power cord required
  • Rugged, chemical-resistant housing
IFT10HP Model
  • High Precision optical instrument with improved 0-10 scale 0.1 scale increments for testing low concentration synthetic cutting and grinding fluids and lubricants
  • 10X focusable eyepiece
  • Coated antireflection glass optical components improve illumination and shadowline contrast
IFT40 Rhino Model
  • Sealed housing and eyepiece; IP67 rated water resistant and dustproof
  • Black polycarbonate housing reduces stray light
  • 0-30 arbitrary scale is divided into 0.5 increments; accuracy to ±0.25%
  • Eyepiece with large depth of focus
  • Built for production floor use; impact resistant to withstand repeated drops from three feet
Warranty
One year
Specifications
Specifications
0 to 30 Brix
Distilled Water
0.5%
Durable Plastic carrying case, sample application pipets, calibration adjustment wrench
1.33 to 1.38
±0.2
Ideal for applications including Metalworking coolants and lubricants, heat treating fluids, hydraulic fluids, firefighting Foam and plating baths
15°C to 38°C
Product Suggestions
Product Suggestions