missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Regular Grade Leather Drivers Gloves

Catalog No. 19071373
Change view
Click to view available options
Size:
Small
Medium
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19071373 Medium
19071371 Small
19071377 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19071373 Supplier Fisherbrand™ Supplier No. 68161SB/M

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gloves provide abrasion resistance and dexterity

  • Ideal for construction, machine operation, utility work, farming, roping, and other general applications
  • Top-grain leather
  • Keystone thumb
  • Launderable

Specifications

Color Natural
Grade Regular
Hand Specific
Material Top Grain Cowhide Leather
Product Type Driver Gloves
Size Medium
Style Standard
For Use With (Application) Construction, Machine Operation, Light Duty Welding, Utility Work, Farming, Roping, and Many Other General Applications
Glove Cuff Style Open
Hem Color Green
Thumb Style Keystone
Length (English) 9.3 in.
Length (Metric) 23.5 cm
Show More Show Less