Learn More
PYREX™ Regular Cyanide Distillation Apparatus Replacement Parts
Catalog No. 0572316D
Change view
Click to view available options
Type:
Dispersion Tube
Inlet Tube
Trap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
0572316D Dispersion Tube
0572316E Trap
0572316A Inlet Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0572316D Supplier PYREX™ Supplier No. 3350DTO
Replacement parts for use with Corning PYREX Regular Cyanide Distillation Apparatus
For Cyanide Distillation Apparatus, see cat. no. 0572316
Compliance
Meets U.S. EPA SW-846 Methods 9010 and 9012 and ASTM™ Method D 2036.
Specifications
For Use With (Application) Pyrex™ cyanide distilling apparatus
Includes Standard Taper Inner Joint 29/42
Material Borosilicate Glass
Type Dispersion Tube