missing translation for 'onlineSavingsMsg'
Learn More

Regorafenib 98.0+%, TCI America™

Catalog No. r0142100mg
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
25 mg
Catalog No. Quantity
R0142100MG 100 mg
R014225MG 25 mg
2 options

Catalog No. R0142100MG

Supplier: TCI America R0142100MG

Only null left

Chemical Identifiers

CAS 755037-03-7
Molecular Formula C21H15ClF4N4O3
Molecular Weight (g/mol) 482.82
MDL Number MFCD16038047
InChI Key FNHKPVJBJVTLMP-UHFFFAOYSA-N
PubChem CID 11167602
ChEBI CHEBI:68647
IUPAC Name 4-[4-[[4-chloro-3-(trifluoromethyl)phenyl]carbamoylamino]-3-fluorophenoxy]-N-methylpyridine-2-carboxamide
SMILES CNC(=O)C1=NC=CC(=C1)OC2=CC(=C(C=C2)NC(=O)NC3=CC(=C(C=C3)Cl)C(F)(F)F)F

Specifications

Melting Point 208°C
Quantity 100 mg
Formula Weight 482.82
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Regorafenib
TSCA No
Recommended Storage Freezer