missing translation for 'onlineSavingsMsg'
Learn More

Regorafenib 98.0+%, TCI America™

Catalog No. p-7135723
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R014225MG 25 mg
R0142100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R014225MG Supplier TCI America Supplier No. R014225MG

Chemical Identifiers

CAS 755037-03-7
Molecular Formula C21H15ClF4N4O3
Molecular Weight (g/mol) 482.82
MDL Number MFCD16038047
InChI Key FNHKPVJBJVTLMP-UHFFFAOYSA-N
PubChem CID 11167602
ChEBI CHEBI:68647
IUPAC Name 4-[4-[[4-chloro-3-(trifluoromethyl)phenyl]carbamoylamino]-3-fluorophenoxy]-N-methylpyridine-2-carboxamide
SMILES CNC(=O)C1=NC=CC(=C1)OC2=CC(=C(C=C2)NC(=O)NC3=CC(=C(C=C3)Cl)C(F)(F)F)F

Specifications

Melting Point 208°C
Quantity 25 mg
Formula Weight 482.82
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Regorafenib
TSCA No
Recommended Storage Freezer