missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Regenerated Cellulose Tangential Flow Filtration Cassette for up to 0.5 L
Description
- Link the cassettes quickly and securely to a suitable pump and reservoirs, right out of the box.
- All-in-one design avoids the need for expensive cassette holders and TFF systems.
- Advanced flow path and fixed occlusion restrictor eliminate the effort that is usually needed for TFF.
- High performance PES or Hydrosart™ RC membranes ensure maximum recoveries.
- Improve safety for you and your sample with no need for flushing or cleaning.
- Cassettes contain 30 to 60% less plastic and are supplied with only the tubing you need.
- Membrane material: Regenerated cellulose
- Recommended feed volume: 100 to 500 mL
Specifications
Specifications
| Filter Type | Ultrafiltration |
| Format | Cassette |
| For Use With (Equipment) | Peristaltic Pump |
| Material | Polycarbonate |
| Material (Membrane) | Regenerated Cellulose |
| MWCO | 2 kDa |
| Sample Volume | 0.1 to 0.5 L |
| Sterility | Non-sterile |
| Type | TFF cassette |
| Volume (Metric) | 0.5 L |