missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Reflectoquant™ Peroxide Test Strips

Catalog No. M1169740001
Change view
Click to view available options
Range:
0.2 to 20mg/L
100 to 1000mg/L
Type:
Test Strip
No. of Tests:
50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range Type No. of Tests
M1169740001 0.2 to 20mg/L Test Strip 50
M1167310001 100 to 1000mg/L Test Strip 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1169740001 Supplier MilliporeSigma™ Supplier No. 1.16974.0001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Tube containing 50 strips, 1 bar-code strip.

For determination of inorganic peroxides in aqueous solutions

  • Enzymatic reaction method ideal for screening
  • Barcode calibration for reliable, quantitative results
  • Low analysis costs

Reclose the tube containing the test strips immediately after use.

For food and water analysis

Specifications

For Use With (Application) For food and water analysis
For Use With (Equipment) Reflectometer
Graduations 0.2 to 20mg/L
No. of Tests 50
Range 0.2 to 20mg/L
Storage Requirements 2°C to 8°C
Test Method Enzymatic Reaction
Type Test Strip