missing translation for 'onlineSavingsMsg'
Learn More

Reference Material of Gasoline Components Certified by The Japan Petroleum Institute, TCI America™

Catalog No. S0429SET
Change view
Click to view available options
Quantity:
Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0429SET Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0429SET Supplier TCI America Supplier No. S04291SET
Only null left

/ Methanol 1.5 vol% level / Ethanol 5.3 vol% level / MTBE 5.2 vol% level / ETBE 5.2 vol% level / Benzene 0.5 vol% level / Toluene 16.2 vol% level / Xylene 10.7 vol% level / Kerosene 3.7 vol% level (10mLx3)

Specifications

UN Number 1203
Quantity Set
Chemical Name or Material Reference Material of Gasoline Components Certified by The Japan Petroleum Institute, / Methanol 1.5